2-Hydroxy-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8780-250M
COBBPN-8780-250MEA 385 EUR
COBBPN-8780-250M
2-Hydroxy-5-pyrimidinboronsäurepinakolester
2-Hydroxy-5-pyrimidinboronsäurepinakolester
Formel: C₁₀H₁₅BN₂O₃
Molecular Weight: 222,05 g/mol
MDL: MFCD09037497
CAS-Nummer: 1073354-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR