2-Amino-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

PN-8792-1G
COBBPN-8792-1GEA 257 EUR
COBBPN-8792-1G
2-Amino-3-pyridinboronsäurepinakolester
2-Amino-3-pyridinboronsäurepinakolester
Formel: C₁₁H₁₇BN₂O₂
Molecular Weight: 220,08 g/mol
MDL: MFCD09260488
CAS-Nummer: 1073354-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR