4-Amino-2-chlorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Amino-2-chlorophenylboronic acid,pinacolester, 4-Amino-2-chlorophenylboronic acid, piancol ester

PN-8814-1G
COBBPN-8814-1GEA 451 EUR
COBBPN-8814-1G
4-Amino-2-chlorphenylboronsäurepinakolester
4-Amino-2-chlorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BClNO₂
Molecular Weight: 253,54 g/mol
MDL: MFCD09864184
CAS-Nummer: 877160-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR