1-Ethyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(1-Ethyl-1H-pyrazol-4yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Ethyl-1H-pyrazole-4-boronic acid Pinacol ester, 1-Ethyl-1H-pyrazole-4-boronic acid, pinacol ester, 1-Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

PN-8817-1G
COBBPN-8817-1GEA 88.3 EUR
COBBPN-8817-1G
1-Ethyl-1H-pyrazol-4-boronsäurepinakolester
1-Ethyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₁H₁₉BN₂O₂
Molecular Weight: 222,0918 g/mol
MDL: MFCD09864188
CAS-Nummer: 847818-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR