1,3-Dimethyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1,3-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1,3-Dimethylpyrazole-4-boronic acid Pinacol ester, 1,3-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 2-(1,3-Dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8819-1G
COBBPN-8819-1GEA 66.3 EUR
COBBPN-8819-1G
1,3-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
1,3-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₁H₁₉BN₂O₂
Molecular Weight: 222,0918 g/mol
MDL: MFCD09864190
CAS-Nummer: 1046832-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR