2-Cyan-5-fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

PN-8821-1G
COBBPN-8821-1GEA 50.9 EUR
COBBPN-8821-1G
2-Cyan-5-fluorphenylboronsäurepinakolester
2-Cyan-5-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₅BFNO₂
Molecular Weight: 247,700 g/mol
MDL: MFCD09878540
CAS-Nummer: 463335-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR