3-Cyan-1-propylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile

PN-8832-1G
COBBPN-8832-1GEA 207 EUR
COBBPN-8832-1G
3-Cyan-1-propylboronsäurepinakolester
3-Cyan-1-propylboronsäurepinakolester
Formel: C₁₀H₁₈BNO₂
Molecular Weight: 195,07 g/mol
MDL: MFCD09953480
CAS-Nummer: 238088-16-9
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR