3-(2-Hydroxyphenyl)propylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8833-1G
COBBPN-8833-1GEA 298 EUR
COBBPN-8833-1G
3-(2-Hydroxyphenyl)propylboronsäurepinakolester
3-(2-Hydroxyphenyl)propylboronsäurepinakolester
Formel: C₁₅H₂₃BO₃
MDL: MFCD09953481
CAS-Nummer: 1073355-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR