4-Iodphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8838-1G
COBBPN-8838-1GEA 0 EUR
COBBPN-8838-1G
4-Iodphenylboronsäurepinakolester
4-Iodphenylboronsäurepinakolester
Formel: C₁₂H₁₆BIO₂
MDL: MFCD09953499
CAS-Nummer: 73852-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR