3-(4-Methoxyphenyl)propylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester

PN-8841-250M
COBBPN-8841-250MEA 451 EUR
COBBPN-8841-250M
3-(4-Methoxyphenyl)propylboronsäurepinakolester
3-(4-Methoxyphenyl)propylboronsäurepinakolester
Formel: C₁₆H₂₅BO₃
MDL: MFCD09953503
CAS-Nummer: 1073371-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR