3-Brom-1-propylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3-Bromopropylboronic acid pinacol ester, 2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-8846-1G
PN-8846-5G
COBBPN-8846-1GEA
127
EUR
COBBPN-8846-1G
COBBPN-8846-5G
3-Brom-1-propylboronsäurepinakolester
3-Brom-1-propylboronsäurepinakolester
Formel:
C₉H₁₈BBrO₂ Molecular Weight: 248.96 g/mol |
MDL:
MFCD10567053 CAS-Nummer: 124215-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...