3-Brom-1-propylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromopropylboronic acid pinacol ester, 2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8846-1G PN-8846-5G
COBBPN-8846-1GEA 127 EUR
COBBPN-8846-1G COBBPN-8846-5G
3-Brom-1-propylboronsäurepinakolester
3-Brom-1-propylboronsäurepinakolester
Formel: C₉H₁₈BBrO₂
Molecular Weight: 248.96 g/mol
MDL: MFCD10567053
CAS-Nummer: 124215-44-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR