4-(Morpholino)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 4-Morpholinophenylboronic acid Pinacol ester, 4-(4-Morpholinyl)benzeneboronic acid pinacol ester, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine

PN-8854-1G
COBBPN-8854-1GEA 121 EUR
COBBPN-8854-1G
4-(Morpholino)phenylboronsäurepinakolester
4-(Morpholino)phenylboronsäurepinakolester
Formel: C₁₆H₂₄BNO₃
Molecular Weight: 289,18 g/mol
MDL: MFCD04112544
CAS-Nummer: 568577-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR