1-Isopentyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-Isopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-(3-Methylbutyl)-1H-pyrazole-4-boronic acid, pinacol ester

PN-8862-1G
COBBPN-8862-1GEA 183 EUR
COBBPN-8862-1G
1-Isopentyl-1H-pyrazol-4-boronsäurepinakolester
1-Isopentyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₄H₂₅BN₂O₂
Molecular Weight: 264,1700 g/mol
MDL: MFCD05663867
CAS-Nummer: 777063-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR