2-Amino-5-methylphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Amino-5-methylphenyboronic acid, pinacol ester, 4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
PN-8880-1G
PN-8880-250M
COBBPN-8880-1GEA
528
EUR
COBBPN-8880-1G
COBBPN-8880-250M
2-Amino-5-methylphenylboronsäurepinakolester
2-Amino-5-methylphenylboronsäurepinakolester
Formel:
C₁₃H₂₀BNO₂ Molecular Weight: 233.12 g/mol |
MDL:
MFCD06795664 CAS-Nummer: 948592-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...