2-Propoxy-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Propoxypyridine-3-boronic acid, pinacol ester, 2-Propoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-8894-1G
COBBPN-8894-1GEA 410 EUR
COBBPN-8894-1G
2-Propoxy-3-pyridinboronsäurepinakolester
2-Propoxy-3-pyridinboronsäurepinakolester
Formel: C₁₄H₂₂BNO₃
Molecular Weight: 263,15 g/mol
MDL: MFCD07781169
CAS-Nummer: 1073371-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR