1-(Phenylsulfonyl)-3-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-(Phenylsulphonyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

PN-8903-1G
COBBPN-8903-1GEA 153 EUR
COBBPN-8903-1G
1-(Phenylsulfonyl)-3-indolboronsäurepinakolester
1-(Phenylsulfonyl)-3-indolboronsäurepinakolester
Formel: C₂₀H₂₂BNO₄S
Molecular Weight: 383,27 g/mol
MDL: MFCD08276778
CAS-Nummer: 870717-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR