3,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 3,5-Dimethylpyrazole-4-boronic acid pinacol ester

PN-8919-1G
COBBPN-8919-1GEA 102 EUR
COBBPN-8919-1G
3,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
3,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₁H₁₉BN₂O₂
Molecular Weight: 222,0918 g/mol
MDL: MFCD05663852
CAS-Nummer: 857530-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR