4-Amino-2-fluorphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-Amino-2-fluorobenzeneboronic acid pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
PN-8921-1G
PN-8921-5G
COBBPN-8921-1GEA
0
EUR
COBBPN-8921-1G
COBBPN-8921-5G
4-Amino-2-fluorphenylboronsäurepinakolester
4-Amino-2-fluorphenylboronsäurepinakolester
Formel:
C₁₂H₁₇BFNO₂ |
MDL:
MFCD09951877 CAS-Nummer: 819057-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...