4-Amino-2-fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Amino-2-fluorobenzeneboronic acid pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

PN-8921-1G PN-8921-5G
COBBPN-8921-1GEA 0 EUR
COBBPN-8921-1G COBBPN-8921-5G
4-Amino-2-fluorphenylboronsäurepinakolester
4-Amino-2-fluorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BFNO₂
MDL: MFCD09951877
CAS-Nummer: 819057-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR