4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8932-5G PN-8932-1G
COBBPN-8932-5GEA 0 EUR
COBBPN-8932-5G COBBPN-8932-1G
4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
Formel: C₁₆H₂₃BO₄
MDL: MFCD08704684
CAS-Nummer: 859169-20-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR