3-Brom-2-fluor-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-2-fluoropyridine-4-boronic acid pinacol ester

PN-9023-1G
COBBPN-9023-1GEA 227 EUR
COBBPN-9023-1G
3-Brom-2-fluor-4-pyridinboronsäurepinakolester
3-Brom-2-fluor-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₄BBrFNO₂
MDL: MFCD08063110
CAS-Nummer: 1150561-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR