4-(Piperidin-1-ylmethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-((Azacyclohex-1-yl)methyl)benzolboronsäure(tetramethylethylen)ester

PN-9089-1G
COBBPN-9089-1GEA 102 EUR
COBBPN-9089-1G
4-(Piperidin-1-ylmethyl)phenylboronsäurepinakolester
4-(Piperidin-1-ylmethyl)phenylboronsäurepinakolester
Formel: C₁₈H₂₈BNO₂
Molecular Weight: 301,23 g/mol
MDL: MFCD08271932
CAS-Nummer: 859833-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR