1H-Benzimidazol-5-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-9095-1G
COBBPN-9095-1GEA 0 EUR
COBBPN-9095-1G
1H-Benzimidazol-5-boronsäurepinakolester
1H-Benzimidazol-5-boronsäurepinakolester
Formel: C₁₃H₁₇BN₂O₂
MDL: MFCD11054041
CAS-Nummer: 1007206-54-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR