2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-5-boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)pyridine
PN-9116-1G
PN-9116-250M
COBBPN-9116-1GEA
0
EUR
COBBPN-9116-1G
COBBPN-9116-250M
2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-5-boronsäurepinakolester
2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-5-boronsäurepinakolester
Formel:
C₁₆H₂₄BNO₄ |
MDL:
MFCD09702390 CAS-Nummer: 910036-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...