2-Methyl-2H-indazol-6-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

PN-9131-1G
COBBPN-9131-1GEA 362 EUR
COBBPN-9131-1G
2-Methyl-2H-indazol-6-boronsäurepinakolester
2-Methyl-2H-indazol-6-boronsäurepinakolester
Formel: C₁₄H₁₉BN₂O₂
Molecular Weight: 258,1239 g/mol
MDL: MFCD11109435
CAS-Nummer: 1204580-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR