2-(Ethoxycarbonyl)phenylboronsäurepropandiolcyclischester

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-(1,3,2-dioxaborinan-2-yl)benzoate

PR-2126-5G PR-2126-1G
COBBPR-2126-5GEA 0 EUR
COBBPR-2126-5G COBBPR-2126-1G
2-(Ethoxycarbonyl)phenylboronsäurepropandiolcyclischester
2-(Ethoxycarbonyl)phenylboronsäurepropandiolcyclischester
Formel: C₁₂H₁₅BO₄
MDL: MFCD06659932
CAS-Nummer: 850567-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR