4-(Methylcarbamoyl)phenylboronsäurepropandiolcyclischester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(1,3,2-Dioxaborinan-2-yl)-N-methylbenzamide

PR-3006-1G PR-3006-5G
COBBPR-3006-1GEA 0 EUR
COBBPR-3006-1G COBBPR-3006-5G
4-(Methylcarbamoyl)phenylboronsäurepropandiolcyclischester
4-(Methylcarbamoyl)phenylboronsäurepropandiolcyclischester
Formel: C₁₁H₁₄BNO₃
MDL: MFCD09801045
CAS-Nummer: 957120-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR