8-Bromchinolin-2(1H)-on
Lieferant: COMBI-BLOCKS
PY-1183-5G
PY-1183-1G
COBBPY-1183-5GEA
543
EUR
COBBPY-1183-5G
COBBPY-1183-1G
8-Bromchinolin-2(1H)-on
8-Bromchinolin-2(1H)-on
Formel:
C₉H₆BrNO Molecular Weight: 224.0540 g/mol |
MDL:
MFCD08741409 CAS-Nummer: 67805-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...