Methyl-6-amino-3,5-dibrompicolinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 6-amino-3,5-dibromopicolinate

PY-1191-1G
COBBPY-1191-1GEA 288 EUR
COBBPY-1191-1G
Methyl-6-amino-3,5-dibrompicolinat
Methyl-6-amino-3,5-dibrompicolinat
Formel: C₇H₆Br₂N₂O₂
Molecular Weight: 309,9400 g/mol
MDL: MFCD13195771
CAS-Nummer: 443956-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR