Methyl-5-fluorpicolinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 5-fluoropicolinate, Methyl 5-fluoropyridine-2-carboxylate
PY-1211-1G
COBBPY-1211-1GEA
165
EUR
COBBPY-1211-1G
Methyl-5-fluorpicolinat
Methyl-5-fluorpicolinat
Formel:
C₇H₆FNO₂ Molecular Weight: 155,1300 g/mol |
MDL:
MFCD11845630 CAS-Nummer: 107504-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...