2,4-Dichlor-5-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2,4-Dichloro-5-picoline

PY-1253-1G
COBBPY-1253-1GEA 41.3 EUR
COBBPY-1253-1G
2,4-Dichlor-5-methylpyridin
2,4-Dichloro-5-methylpyridine
Formel: C₆H₅Cl₂N
Molecular Weight: 162,02 g/mol
MDL: MFCD08704813
CAS-Nummer: 56961-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR