5-Brom-2-methoxy-4-methyl-3-nitropyridin
Lieferant: COMBI-BLOCKS
PY-1276-1G
PY-1276-25G
PY-1276-5G
COBBPY-1276-1GEA
65.8
EUR
COBBPY-1276-1G
COBBPY-1276-25G
COBBPY-1276-5G
5-Brom-2-methoxy-4-methyl-3-nitropyridin
5-Brom-2-methoxy-4-methyl-3-nitropyridin
Formel:
C₇H₇BrN₂O₃ Molecular Weight: 247.0500 g/mol |
MDL:
MFCD06659504 CAS-Nummer: 884495-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...