3-Brom-5-chlorpyridin-N-oxid

Lieferant: COMBI-BLOCKS

PY-1357-1G
COBBPY-1357-1GEA 204 EUR
COBBPY-1357-1G
3-Brom-5-chlorpyridin-N-oxid
3-Brom-5-chlorpyridin-N-oxid
Formel: C₅H₃BrClNO
MDL: MFCD16036154
CAS-Nummer: 1221793-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR