3-Brom-5-fluorpyridin-N-oxid

Lieferant: COMBI-BLOCKS

PY-1358-5G
COBBPY-1358-5GEA 0 EUR
COBBPY-1358-5G
3-Brom-5-fluorpyridin-N-oxid
3-Brom-5-fluorpyridin-N-oxid
Formel: C₅H₃BrFNO
MDL: MFCD16036155
CAS-Nummer: 1221793-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR