3-Chlor-5-fluorpyridin-N-oxid

Lieferant: COMBI-BLOCKS

PY-1360-1G
COBBPY-1360-1GEA 363 EUR
COBBPY-1360-1G
3-Chlor-5-fluorpyridin-N-oxid
3-Chlor-5-fluorpyridin-N-oxid
Formel: C₅H₃ClFNO
MDL: MFCD16036157
CAS-Nummer: 1221793-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR