(3-Fluor-2-pyridinyl)methanol
Lieferant: COMBI-BLOCKS
Synonyme:
3-Fluoro-2-(hydroxymethyl)pyridine, (3-Fluoropyrid-2-yl)methanol
PY-1451-1G
COBBPY-1451-1GEA
76.1
EUR
COBBPY-1451-1G
(3-Fluor-2-pyridinyl)methanol
(3-Fluor-2-pyridinyl)methanol
Formel:
C₆H₆FNO Molecular Weight: 127,1200 g/mol |
MDL:
MFCD09025826 CAS-Nummer: 31181-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...