5-Methylnicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Methyl-3-pyridincarbonsäure, 5-Methylpyridin-3-carbonsäure

PY-1457-100G PY-1457-25G PY-1457-5G
COBBPY-1457-100GEA 0 EUR
COBBPY-1457-100G COBBPY-1457-25G COBBPY-1457-5G
5-Methylnicotinsäure
5-Methylnicotinsäure
Formel: C₇H₇NO₂
MDL: MFCD00829036
CAS-Nummer: 3222-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR