5-Brom-2-methoxy-3-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-methoxy-3-methylpyridine

PY-1464-25G
COBBPY-1464-25GEA 456 EUR
COBBPY-1464-25G
5-Brom-2-methoxy-3-methylpyridin
5-Brom-2-methoxy-3-methylpyridin
Formel: C₇H₈BrNO
Molecular Weight: 202,05 g/mol
MDL: MFCD07375124
CAS-Nummer: 760207-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR