2-Amino-5-brom-3-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-3-(trifluoromethyl)pyridin-2-amine

PY-1517-5G
COBBPY-1517-5GEA 221 EUR
COBBPY-1517-5G
2-Amino-5-brom-3-(trifluormethyl)pyridin
2-Amino-5-bromo-3-(trifluoromethyl)pyridine
Formel: C₆H₄BrF₃N₂
Molecular Weight: 241.01 g/mol
MDL: MFCD10758088
CAS-Nummer: 79456-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR