2-Amino-7-bromchinazolin

Lieferant: COMBI-BLOCKS

Synonyme: 7-Bromoquinazolin-2-amine

PY-1754-1G
COBBPY-1754-1GEA 247 EUR
COBBPY-1754-1G
2-Amino-7-bromchinazolin
2-Amino-7-bromoquinazoline
Formel: C₈H₆BrN₃
Molecular Weight: 224,0600 g/mol
MDL: MFCD09027698
CAS-Nummer: 190274-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR