3,5-Dibrom-2-chlorpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-3,5-dibromopyridine

PY-1864-25G
COBBPY-1864-25GEA 221 EUR
COBBPY-1864-25G
3,5-Dibrom-2-chlorpyridin
3,5-Dibrom-2-chlorpyridin
Formel: C₅H₂Br₂ClN
Molecular Weight: 271,34 g/mol
MDL: MFCD00233991
CAS-Nummer: 40360-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR