4-Brom-2,6-dichlorpyridin

Lieferant: COMBI-BLOCKS

PY-1959-5G
COBBPY-1959-5GEA 307 EUR
COBBPY-1959-5G
4-Brom-2,6-dichlorpyridin
4-Bromo-2,6-dichloropyridine
Formel: C₅H₂BrCl₂N
Molecular Weight: 226,8900 g/mol
MDL: MFCD11848646
CAS-Nummer: 98027-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR