N-Benzyl-4-(benzylamino)picolinamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Benzylamino)-N-benzylpyridine-2-carboxamide

PY-7020-1G PY-7020-5G
COBBPY-7020-1GEA 149 EUR
COBBPY-7020-1G COBBPY-7020-5G
N-Benzyl-4-(benzylamino)picolinamid
N-Benzyl-4-(benzylamino)picolinamid
Formel: C₂₀H₁₉N₃O
MDL: MFCD09027296
CAS-Nummer: 913836-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR