5-Brom-2-fluorpyridin

Lieferant: COMBI-BLOCKS

PY-7039-100G PY-7039-25G PY-7039-5G
COBBPY-7039-100GEA 205 EUR
COBBPY-7039-100G COBBPY-7039-25G COBBPY-7039-5G
5-Brom-2-fluorpyridin
5-Brom-2-fluorpyridin
Formel: C₅H₃BrFN
Molecular Weight: 175.99 g/mol
Siedepunkt: 63 °C (15 mmHg)
MDL: MFCD01863742
CAS-Nummer: 766-11-0
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR