3-Chlor-2-hydrazinopyridin

Lieferant: COMBI-BLOCKS

PY-7086-1G PY-7086-5G
COBBPY-7086-1GEA 0 EUR
COBBPY-7086-1G COBBPY-7086-5G
3-Chlor-2-hydrazinopyridin
3-Chlor-2-hydrazinopyridin
Formel: C₅H₆ClN₃
Molecular Weight: 143.58 g/mol
MDL: MFCD01936479
CAS-Nummer: 22841-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR