3,6-Dibrom-2-picolin

Lieferant: COMBI-BLOCKS

Synonyme: 3,6-Dibrom-2-methylpyridin

PY-7092-1G PY-7092-25G PY-7092-5G
COBBPY-7092-1GEA 0 EUR
COBBPY-7092-1G COBBPY-7092-25G COBBPY-7092-5G
3,6-Dibrom-2-picolin
3,6-Dibrom-2-picolin
Formel: C₆H₅Br₂N
Molecular Weight: 250.92 g/mol
MDL: MFCD06254589
CAS-Nummer: 39919-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR