4-Brom-2-chlor-3-iodpyridin

Lieferant: COMBI-BLOCKS

PY-7095-1G
COBBPY-7095-1GEA 303 EUR
COBBPY-7095-1G
4-Brom-2-chlor-3-iodpyridin
4-Bromo-2-chloro-3-iodopyridine
Formel: C₅H₂BrClIN
Molecular Weight: 318,34 g/mol
MDL: MFCD08060939
CAS-Nummer: 916203-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR