8-Chlortetrazolo[1,5-a]pyridin

Lieferant: COMBI-BLOCKS

PY-7099-1G
COBBPY-7099-1GEA 149 EUR
COBBPY-7099-1G
8-Chlortetrazolo[1,5-a]pyridin
8-Chlortetrazolo[1,5-a]pyridin
Formel: C₅H₃ClN₄
MDL: MFCD00490111
CAS-Nummer: 40971-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR