6-Brom-3-(ethylamino)-[1,2,4]triazolo[4,3-a]pyridin

Lieferant: COMBI-BLOCKS

PY-7110-1G
COBBPY-7110-1GEA 497 EUR
COBBPY-7110-1G
6-Brom-3-(ethylamino)-[1,2,4]triazolo[4,3-a]pyridin
6-Brom-3-(ethylamino)-[1,2,4]triazolo[4,3-a]pyridin
Formel: C₈H₉BrN₄
MDL: MFCD09801048
CAS-Nummer: 951884-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR